missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nogo-A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
178.00 € - 512.00 €
Specifications
| Antigen | Nogo-A |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18693156
|
Novus Biologicals
NBP2-41071-0.025mg |
0.025 mg |
178.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18124038
|
Novus Biologicals
NBP2-41071 |
0.1 mg |
512.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nogo-A Polyclonal specifically detects Nogo-A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Nogo-A | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| RTN4 | |
| IgG | |
| Affinity Purified | |
| ∼180 kDa |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| 57142 | |
| Antibody was raised against a 19 amino acid synthetic peptide from near the center of human NogoA. The immunogen is located within amino acids 200 - 250 of NogoA. Amino Acid Squence: DLKEQPGNTISAGQEDFPS | |
| Primary | |
| At least five isoforms of Nogo are known to exist; this antibody is specific for NogoA and NogoE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title