missing translation for 'onlineSavingsMsg'
Learn More

Nogo Receptor/NgR Antibody (SR2112), Novus Biologicals™

Product Code. 18622625 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18622625 100 μg 100µL
18673045 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18622625 Supplier Novus Biologicals Supplier No. NBP322117100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Nogo Receptor/NgR Monoclonal antibody specifically detects Nogo Receptor/NgR in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nogo Receptor/NgR
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone SR2112
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, Immunoprecipitation 1:50
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias NgR, Nogo receptor, Nogo-66 receptor, NOGORUNQ330/PRO526, reticulon 4 receptor, reticulon-4 receptor
Host Species Rabbit
Immunogen A synthesized peptide derived from human Nogo Receptor/NgR (Uniprot #: Q9BZR6)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 65078
Target Species Human, Mouse
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.