missing translation for 'onlineSavingsMsg'
Learn More

NOP14 Antibody [Alexa Fluor« 750], Novus Biologicals Biologicals™

Product Code. 30489304 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30489304 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30489304 Supplier Novus Biologicals Supplier No. NBP335129AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NOP14 Polyclonal antibody specifically detects NOP14 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOP14
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias C4orf9, chromosome 4 open reading frame 9, NOL14nucleolar protein 14 homolog (yeast), NOP14 homolog, NOP14 nucleolar protein homolog (yeast), Nucleolar complex protein 14, nucleolar protein 14, nucleolar protein 14 homolog, probable nucleolar complex protein 14, RES425, RES4-25
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human NOP14 (NP_001278908.1).,, Sequence:, MAKAKKVGARRKASGAPAGARGGPAKANSNPFEVKVNRQKFQILGRKTRHDVGLPGVSRARALRKRTQTLLKEYKERDKSNVFRDKRFGEYNSNMSPEEKMMKRFALEQQRHHEKKSIYNLNEDEELTHYGQSLADIEKHNDIVDSDSDAEDRGTLSAELTAAHFGGGGGLLHKKTQQEGEEREKPKSRKELIEELIAKSKQEKRERQAQREDALELTEKLDQDWKEIQTLLSHKTPKSE
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8602
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.