missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Norclozapine.HCl, Lipomed
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Norclozapine.HCl
Specifications
Specifications
| Chemical Name or Material | Norclozapine Hydrochloride |
| CAS | 2741767-92-8 |
| Analyte or Component Names | Norclozapine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H17 Cl N4 . Cl H |
| Synonym | 5 H-Dibenzo[b,e][1,4 ]diazepine, 8-chloro-11-(1-piperazinyl)-, hydrochloride (1:1) (ACI), 8-Chloro-11-piperazin-1-yl-5 H-dibenzo[b,e][1,4 ]diazepine Hydrochloride, Norclozapine HCl |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.Clc1ccc2Nc3ccccc3C(=Nc2c1)N4CCNCC4 |
| IUPAC Name | 3-chloro-6-piperazin-1-yl-11 H-benzo[b][1,4 ]benzodiazepine;hydrochloride |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction