missing translation for 'onlineSavingsMsg'
Learn More

Nortriptyline Hydrochloride, TRC

Product Code. 30138652
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138652 100 mg 100mg
30135249 1 g 1g
17930685 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138652 Supplier Toronto Research Chemicals Supplier No. TRCN837000100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 894-71-3
Molecular Formula C19 H21 N . Cl H
Molecular Weight (g/mol) 299.84
Synonym Nortriptyline Hydrochloride, 3-(10,11-Dihydro-5H-dibenzo[a,d][7]annulen-5-ylidene)-N-methylpropan-1-amine Hydrochloride, Amitriptyline Hydrochloride Imp. C (EP) as Hydrochloride, 1-Propanamine, 3-(10,11-dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N-methyl-, hydrochloride (1:1), 1-Propanamine, 3-(10,11-dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N-methyl-, hydrochloride (9CI), 5H-Dibenzo[a,d]cycloheptene-δ5,γ-propylamine, 10,11-dihydro-N-methyl-, hydrochloride (8CI), 10,11-Dihydro-N-methyl-5H-dibenzo[a,d]cycloheptene-δ5,γ-propylamine hydrochloride, 38489, 5-(3-Methylaminopropylidene)-10,11-dihydro-5H-dibenzo[a,d]cycloheptene hydrochloride, 5-[3-(Methylamino)propylidene]dibenzo[a,e]cyclohepta[1,5]diene hydrochloride, Acetexa, Allegron, Altilev, Aventyl hydrochloride, Desmethylamitriptyline hydrochloride, Noramitriptyline hydrochloride, Norfenazin, Noritren, Nortrilen, Nortryptilinium hydrochloride, Norzepine, Pamelor, Psychostyl, Sensival, Vividyl
IUPAC Name 3-(5,6-dihydrodibenzo[2,1-b:1',2'-e][7]annulen-11-ylidene)-N-methylpropan-1-amine;hydrochloride
SMILES Cl.CNCCC=C1c2ccccc2CCc3ccccc13

Specifications

Shipping Condition Room Temperature
InChI Formula InChI=1S/C19H21N.ClH/c1-20-14-6-11-19-17-9-4-2-7-15(17)12-13-16-8-3-5-10-18(16)19;/h2-5,7-11,20H,6,12-14H2,1H3;1H
Industry Type Pharmaceutical
Quantity 100 mg
Formula Weight 299.1441
Percent Purity >95%
Chemical Name or Material Nortriptyline hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.