missing translation for 'onlineSavingsMsg'
Learn More

NOX1 Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. 18698741 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18698741 0.02 mL 0.02mL
18604892 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18698741 Supplier Novus Biologicals Supplier No. NBP2939670.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NOX1 Polyclonal antibody specifically detects NOX1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOX1
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:500, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias EC 1.6.3, GP91-2, Mitogenic oxidase 1, MOX-1, MOX1NADH/NADPH mitogenic oxidase subunit P65-MOX, NADPH oxidase 1, NADPH oxidase homolog-1, NOH1mitogenic oxidase (pyridine nucleotide-dependent superoxide-generating), NOH-1NADPH oxidase 1 variant NOH-1L, NOX-1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200-300 of human NOX1 (NP_008983.2). YFEVFWYTHHLFIFYILGLGIHGIGGIVRGQTEESMNESHPRKCAESFEMWDDRDSHCRRPKFEGHPPESWKWILAPVILYICERILRFYRSQQKVVITKV
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Direct Reversal of DNA Damage, Endocrinology, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 27035
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.