missing translation for 'onlineSavingsMsg'
Learn More

NOXO1 Antibody, Novus Biologicals™

Product Code. p-200057952 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18632009 25 μL 25µL
18652288 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18632009 Supplier Novus Biologicals Supplier No. NBP24966325ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NOXO1 Polyclonal antibody specifically detects NOXO1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOXO1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2), 40% Glycerol
Gene Alias MGC20258, NADPH oxidase organizer 1, Nox organizer 1, Nox-organizing protein 1, P41NOX, P41NOXA, P41NOXB, P41NOXC, regulatory protein P41NOX, SH3 and PX domain-containing protein 5, SH3PXD5NADPH oxidase regulatory protein, SNX28
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: LLETYSRRLLATAERVARSPTITGFFAPQPLDLE
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cytokine Research, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 124056
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.