missing translation for 'onlineSavingsMsg'
Learn More

NPL Antibody - BSA Free, Novus Biologicals™

Product Code. 30558854 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30558854 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30558854 Supplier Novus Biologicals Supplier No. NBP347312

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NPL Polyclonal antibody specifically detects NPL in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPL
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA, Immunocytochemistry/ Immunofluorescence 1:20-1:200
Formulation PBS, 50% Glycerol, pH (7.3)
Gene Alias C112, C1orf13MGC61869, chromosome 1 open reading frame 13, DHDPS1, dihydrodipicolinate synthetase homolog 1, EC 4.1.3.3, MGC149582, N-acetylneuraminate lyase, N-acetylneuraminate pyruvate lyase (dihydrodipicolinate synthase), N-acetylneuraminate pyruvate-lyase, N-acetylneuraminic acid aldolase, NAL, NALase, NPL1, Sialate lyase, Sialate-pyruvate lyase, Sialic acid aldolase, Sialic acid lyase
Host Species Rabbit
Immunogen N-acetylneuraminate pyruvate lyase(dihydrodipicolinate synthase). (Uniprot# Q9BXD5)
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 80896
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.