missing translation for 'onlineSavingsMsg'
Learn More

Nuclear Factor Erythroid 2 Related Factor 1 Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. 18627160 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.01mL
0.02mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18627160 0.02 mL 0.02mL
18640751 0.1 mL 0.01mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18627160 Supplier Novus Biologicals Supplier No. NBP2937920.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Nuclear Factor Erythroid 2 Related Factor 1 Polyclonal antibody specifically detects Nuclear Factor Erythroid 2 Related Factor 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nuclear Factor Erythroid 2 Related Factor 1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias FLJ00380, HBZ17, LCR-F1, Locus control region-factor 1, NF-E2-related factor 1, NFE2-related factor 1, NRF1TCF11, nuclear factor (erythroid-derived 2)-like 1, nuclear factor erythroid 2-related factor 1, Nuclear factor, erythroid derived 2, like 1, TCF-11, Transcription factor 11, transcription factor 11 (basic leucine zipper type), Transcription factor HBZ17, Transcription factor LCR-F1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 515-772 of human NFE2L1 (NP_003195.1). SSSFSEEGAVGYSSDSETLDLEEAEGAVGYQPEYSKFCRMSYQDPAQLSCLPYLEHVGHNHTYNMAPSALDSADLPPPSALKKGSKEKQADFLDKQMSRDEHRARAMKIPFTNDKIINLPVEEFNELLSKYQLSEAQLSLIRDIRRRGKNKMAAQNCRKRKLDTILNLERDVEDLQRDKARLLREKVEFLRSLRQMKQKVQSLYQEVFGRLRDENGRPYSPSQYALQYAGDGSVLLIPRTMADQQARRQERKPKDRRK
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Cell Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4779
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.