missing translation for 'onlineSavingsMsg'
Learn More

Nuclear Factor Erythroid 2 Related Factor 1 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-93792-0.1ml

397.80 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


 View more versions of this product

Product Code. 18640751

  • 470.00 € / 0.01mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Nuclear Factor Erythroid 2 Related Factor 1 Polyclonal antibody specifically detects Nuclear Factor Erythroid 2 Related Factor 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nuclear Factor Erythroid 2 Related Factor 1
Polyclonal
Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin
FLJ00380, HBZ17, LCR-F1, Locus control region-factor 1, NF-E2-related factor 1, NFE2-related factor 1, NRF1TCF11, nuclear factor (erythroid-derived 2)-like 1, nuclear factor erythroid 2-related factor 1, Nuclear factor, erythroid derived 2, like 1, TCF-11, Transcription factor 11, transcription factor 11 (basic leucine zipper type), Transcription factor HBZ17, Transcription factor LCR-F1
Recombinant fusion protein containing a sequence corresponding to amino acids 515-772 of human NFE2L1 (NP_003195.1). SSSFSEEGAVGYSSDSETLDLEEAEGAVGYQPEYSKFCRMSYQDPAQLSCLPYLEHVGHNHTYNMAPSALDSADLPPPSALKKGSKEKQADFLDKQMSRDEHRARAMKIPFTNDKIINLPVEEFNELLSKYQLSEAQLSLIRDIRRRGKNKMAAQNCRKRKLDTILNLERDVEDLQRDKARLLREKVEFLRSLRQMKQKVQSLYQEVFGRLRDENGRPYSPSQYALQYAGDGSVLLIPRTMADQQARRQERKPKDRRK
0.1 mL
Cell Biology, Immunology
4779
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.