missing translation for 'onlineSavingsMsg'
Learn More

NUP43 Antibody, R&D Systems™

Product Code. 30046557 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30046557 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30046557 Supplier Novus Biologicals Supplier No. NBP329504
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

NUP43 Polyclonal specifically detects NUP43 in Human, Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP43
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Tris-citrate/phosphate buffer, pH 7 to 8
Gene Alias bA350J20.1, FLJ13287, nucleoporin 43kDa, nucleoporin Nup43, Nup107-160 subcomplex subunit Nup43, p42
Host Species Rabbit
Immunogen Between 330 and 380
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 348995
Target Species Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.