missing translation for 'onlineSavingsMsg'
Learn More

NUP93 Antibody - Azide and BSA Free, R&D Systems™

Product Code. 18607802 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18607802 0.02 mL 0.02mL
18651412 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18607802 Supplier Novus Biologicals Supplier No. NBP2948880.02ml
 Restricted Item
Estimated Shipment: 03-12-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

NUP93 Polyclonal specifically detects NUP93 in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen NUP93
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias KIAA0095Nucleoporin Nup93,93 kDa nucleoporin, MGC21106, nuclear pore complex protein Nup93, nucleoporin 93kDa
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 670-819 of human NUP93 (NP_055484.3). IAERYRAQGISANKFVDSTFYLLLDLITFFDEYHSGHIDRAFDIIERLKLVPLNQESVEERVAAFRNFSDEIRHNLSEVLLATMNILFTQFKRLKGTSPSSSSRPQRVIEDRDSQLRSQARTLITFAGMIPYRTSGDTNARLVQMEVLMN
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9688
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.