missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NY-REN-50 antigen Polyclonal antibody specifically detects NY-REN-50 antigen in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | NY-REN-50 antigen |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formulation | PBS, pH7.0 |
| Gene Alias | C14orf131, chromosome 14 open reading frame 131, FLJ11132, Renal carcinoma antigen NY-REN-50, zinc finger protein 839 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human NY-REN-50 antigen fragment |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?