missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 227.0°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5 to 102.5% (Argentometry) |
| Packaging | Glass Bottle |
| Linear Formula | C6F5CH2ONH2·HCl |
| Quantity | 1 g |
| Solubility Information | Solubility in water: soluble. |
| Formula Weight | 249.57 |
| Percent Purity | 98% |
| Chemical Name or Material | O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride |
Safety and Handling
Product Identifier
- o-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction