missing translation for 'onlineSavingsMsg'
Learn More

O-Desmethylsulpiride 1.0 mg/ml in Dimethyl Sulfoxide, LoGiCal

Product Code. 30462618
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30462618

Brand: LoGiCal LGCAMP0051.0729

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

O-Desmethylsulpiride 1.0 mg/ml in Dimethyl Sulfoxide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O-Desmethylsulpiride
CAS 67381-52-6
Analyte or Component Names O-Desmethylsulpiride
Shipping Condition Ice Pack (-20°C)
Molecular Formula C14 H21 N3 O4 S
Certifications/Compliance ISO 17034
Synonym (RS)-N-[(1-Ethylpyrrolidin-2-yl)methyl]-2-hydroxy-5-sulphamoylbenzamide, Sulpiride Imp. G (EP), O-Desmethylsulpiride, Sulpiride Impurity G
InChI Formula InChI=1 S/C14H21N3O4S/c1-2-17-7-3-4-10(17)9-16-14(19)12-8-11(22(15,20)21)5-6-13(12)18/h5-6,8,10,18 H,2-4,7,9H2,1H3,(H,16,19)(H2,15,20,21)
SMILES CCN1CCCC1CNC(=O)c2cc(ccc2O)S(=O)(=O)N
IUPAC Name N-[(1-ethylpyrrolidin-2-yl)methyl]-2-hydroxy-5-sulfamoylbenzamide
Solvent or Matrix Dimethyl Sulfoxide
Industry Type Forensics
Molecular Weight (g/mol) 327.4
Quantity 1 mL
Formula Weight 327.1253 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.