missing translation for 'onlineSavingsMsg'
Learn More

O-((Perfluorophenyl)methyl)hydroxylamine Hydrochloride, TRC

Product Code. 30003309
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003309 100 mg 100mg
30138325 250 mg 250mg
30134045 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003309 Supplier Toronto Research Chemicals Supplier No. TRCP227330100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57981-02-9
Formula Weight 248.998
InChI Formula InChI=1S/C7H4F5NO.ClH/c8-3-2(1-14-13)4(9)6(11)7(12)5(3)10;/h1,13H2;1H
IUPAC Name O-[(2,3,4,5,6-pentafluorophenyl)methyl]hydroxylamine;hydrochloride
Molecular Formula C7 H4 F5 N O . Cl H
Molecular Weight (g/mol) 249.566
SMILES Cl.NOCc1c(F)c(F)c(F)c(F)c1F
Synonym Hydroxylamine, O-[(2,3,4,5,6-pentafluorophenyl)methyl]-, hydrochloride (1:1) (ACI), Hydroxylamine, O-[(pentafluorophenyl)methyl]-, hydrochloride (9CI), ((Perfluorophenyl)methoxy)amine hydrochloride, 2,3,4,5,6-Pentafluorobenzyloxyamine hydrochloride, N-[(2
Chemical Name or Material O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.