missing translation for 'onlineSavingsMsg'
Learn More

OATP1B3/SLCO1B3/OATP8 Antibody (CL3770) [CoraFluor™ 1], Novus Biologicals™

Product Code. 30297277 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30297277 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30297277 Supplier Novus Biologicals Supplier No. NBP261636CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

OATP1B3/SLCO1B3/OATP8 Monoclonal antibody specifically detects OATP1B3/SLCO1B3/OATP8 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen OATP1B3/SLCO1B3/OATP8
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL3770
Conjugate CoraFluor 1
Formulation PBS
Gene Alias Liver-specific organic anion transporter 2, liver-specific organic anion transporter 3TM13, LST2, LST-2, LST3, OATP1B3LST-3TM13, OATP8, OATP-8, Organic anion transporter 8, organic anion transporter LST-3c, Organic anion-transporting polypeptide 8, SLC21A8, solute carrier family 21 (organic anion transporter), member 8, Solute carrier family 21 member 8, solute carrier organic anion transporter family member 1B3, solute carrier organic anion transporter family, member 1B3
Host Species Mouse
Immunogen This antibody was developed using a recombinant protein derived from Q9NPD5, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 28234
Target Species Human
Content And Storage Store at 4⪚C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.