missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Occludin Monoclonal antibody specifically detects Occludin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Occludin |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | rOCLN/8525 |
| Conjugate | DyLight 594 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | BLCPMG, occludin |
| Host Species | Mouse |
| Immunogen | Recombinant human Occludin fragment around aa 282-415 (Exact sequence is proprietary) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?