missing translation for 'onlineSavingsMsg'
Learn More

Octahydrocyclopenta[c]pyrrole Hydrochloride, Mikromol

Product Code. 30461027
Change view
Click to view available options
Quantity:
25 mg
4 x 25 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30461027 4 x 25 mg 100mg
30051495 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30461027 Supplier Mikromol Supplier No. MM0325.1000254AMP
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Octahydrocyclopenta[c]pyrrole Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Octahydrocyclopenta[c]pyrrole Hydrochloride
CAS 112626-50-3
Analyte or Component Names Octahydrocyclopenta[c]pyrrole Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C7 H13 N . Cl H
Certifications/Compliance ISO 9001
Synonym Cyclopenta[c]pyrrole, octahydro-, hydrochloride (1:1), Cyclopenta[c]pyrrole, octahydro-, hydrochloride (6CI,9CI), Octahydrocyclopenta[c]pyrrole Hydrochloride
InChI Formula InChI=1S/C7H13N.ClH/c1-2-6-4-8-5-7(6)3-1;/h6-8H,1-5H2;1H
Solution Type Neat
SMILES Cl.C1CC2CNCC2C1
IUPAC Name 1,2,3,3a,4,5,6,6a-octahydrocyclopenta[c]pyrrole;hydrochloride
Molecular Weight (g/mol) 147.65
Quantity 4 x 25 mg
Formula Weight 147.0815 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.