missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olsalazine Sodium, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Olsalazine sodium |
| CAS | 6054-98-4 |
| Analyte or Component Names | Olsalazine sodium |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H8 N2 O6 . 2 Na |
| Certifications/Compliance | ISO 9001 |
| Synonym | Olsalazine sodium, Disodium 3,3'-diazenediylbis(6-hydroxybenzoate), Benzoic acid, 3,3'-(1,2-diazenediyl)bis[6-hydroxy-, sodium salt (1:2), Benzoic acid, 3,3'-azobis[6-hydroxy-, disodium salt (9CI), C.I. Mordant Yellow 5 (7CI), C.I. Mordant Yellow 5, disodium salt (8CI), 3,3'-Azobis[6-hydroxybenzoic acid] disodium salt, Acid Chrome Flavine R, Alizarol Flavine A, Azodisal sodium, C.I. 14130, Chromaven Yellow FA, Chrome Fast Flavine A, Chrome Fast Flavine J, Chrome Yellow A extra, Chrome Yellow AS, Curolite Yellow 3R, Diamond Chrome Yellow A ex, Dipentum, Disodium 3,3'-azobis(6-hydroxybenzoate), Disodium azodisalicylate, Eriochromal Flavine A, Eriochrome Flavine A, Fenakrom Yellow 3G, Metachrome Yellow A extra, Monochrome Yellow A ex, Mordant Yellow 5, Nutrachrome Yellow NYA, Olsalazine Disodium, Omega Chrome Aurine GL, Ph CJ 91B, Pontachrome Flavine A, Salicine Chrome Yellow A Extra, Sodium azodisalicylate, Solochrome Flavine G, Synchromate Flavine A, Telon Chrome Yellow T, Tertrochrome Yellow A |
| InChI Formula | InChI=1S/C14H10N2O6.2Na/c17-11-3-1-7(5-9(11)13(19)20)15-16-8-2-4-12(18)10(6-8)14(21)22;;/h1-6,17-18H,(H,19,20)(H,21,22);;/q;2*+1/p-2/b16-15+;; |
| Solution Type | Neat |
| SMILES | [Na+].[Na+].Oc1ccc(cc1C(=O)[O-])N=Nc2ccc(O)c(c2)C(=O)[O-] |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?