missing translation for 'onlineSavingsMsg'
Learn More

Omeprazole, Mikromol™

Product Code. 17950736
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17950736 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17950736 Supplier Mikromol Supplier No. MM0095.000250
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Omeprazole
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Omeprazole
CAS 73590-58-6
Analyte or Component Names Omeprazole
Shipping Condition Room Temperature
Molecular Formula C17 H19 N3 O3 S
Certifications/Compliance ISO 17034
Synonym 1H-Benzimidazole, 5-methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl]- (9CI), 2-[[(3,5-Dimethyl-4-methoxy-2-pyridyl)methyl]sulfinyl]-5-methoxy-1H-benzimidazole, 5-Methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridyl)methyl]sulfinyl]-1H-benzimidazole, Acidex, Antra, Antra MUPS, Audazol, Aulcer, Belmazol, Ceprandal, Desec, Dizprazol, Dudencer, Elgam, Emeproton, Epirazole, Gastrimut, GastroGard, Gastroloc, Gastrozole, Gibancer, H 168/68, Indurgan, Inhibitron, Inhipump, Logastric, Lomac, Losec, Mepral, Miol, Miracid, Mopral, OMEP, OMP, OMZ, Ocid, Omapren, Omebeta 20, Omed, Omedar, Omepradex, Omepral, Omeprazen, Omeprazole, Omeprazon, Omepril, Omezol, Omezzol, Omid, Omisec, Omizac, Ompanyt, Oprax, Opraz, Ozoken, Parizac, Pepticum, Peptilcer, Prazidec, Prilosec, Prysma, Ramezol, Roweprazol, Sanamidol, Secrepina, Ulceral, Ulcesep, Ulcometion, Ulcozol, Ulsen, Ultop, Ulzone, Zefxon, Zimor, Zoltum, Chlorphenamine Maleate Imp. C (EP), Chlorphenamine Imp. C (EP), Chlorpheniramine USP Related Compound C, Chlorpheniramine USP RC C, (+/-)-Omeprazole
InChI Formula InChI=1S/C17H19N3O3S/c1-10-8-18-15(11(2)16(10)23-4)9-24(21)17-19-13-6-5-12(22-3)7-14(13)20-17/h5-8H,9H2,1-4H3,(H,19,20)
SMILES COc1ccc2[nH]c(nc2c1)S(=O)Cc3ncc(C)c(OC)c3C
IUPAC Name 5-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]-1H-benzimidazole
Molecular Weight (g/mol) 345.42
Quantity 250 mg
Formula Weight 345.1147 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.