missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Orexin R1/HCRTR1 Antibody - N-terminal, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals™ NBP3-30878-25ug

 View more versions of this product

Product Code. 30066814

  • 213.00 € / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Orexin R1/HCRTR1 Polyclonal antibody specifically detects Orexin R1/HCRTR1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Orexin R1/HCRTR1
Polyclonal
Phosphate buffered saline
Rabbit
Antigen Affinity-purified
RUO
Primary
Human
Purified
ELISA
Unconjugated
hypocretin (orexin) receptor 1, hypocretin receptor 1, Hypocretin receptor type 1, hypocretin receptor-1, orexin receptor 1, orexin receptor type 1, orexin receptor-1, ox-1-R, Ox1-R, OX1R
Synthetic peptide made to the N terminal of human Orexin R1/HCRTR1
25 μg
Cancer, GPCR, Neuronal Cell Markers, Neuroscience
3061
Store at -20°C. Avoid freeze/thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.