missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Orexin R1/HCRTR1 Antibody - N-terminal, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00 € - 508.00 €
Specifications
| Antigen | Orexin R1/HCRTR1 |
|---|---|
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067200
|
Novus Biologicals™
NBP3-30878-100ug |
100 μg |
508.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066814
|
Novus Biologicals™
NBP3-30878-25ug |
25 μg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Orexin R1/HCRTR1 Polyclonal antibody specifically detects Orexin R1/HCRTR1 in Human samples. It is validated for ELISASpecifications
| Orexin R1/HCRTR1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| hypocretin (orexin) receptor 1, hypocretin receptor 1, Hypocretin receptor type 1, hypocretin receptor-1, orexin receptor 1, orexin receptor type 1, orexin receptor-1, ox-1-R, Ox1-R, OX1R | |
| Synthetic peptide made to the N terminal of human Orexin R1/HCRTR1 | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Cancer, GPCR, Neuronal Cell Markers, Neuroscience | |
| Phosphate buffered saline | |
| 3061 | |
| IgG | |
| Antigen Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title