missing translation for 'onlineSavingsMsg'
Learn More

Orotic Acid, Mikromol™

Product Code. 17941446
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17941446 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17941446 Supplier Mikromol Supplier No. MM0881.000250
 Restricted Item
Estimated Shipment: 20-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Orotic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Orotic Acid
CAS 65-86-1
Analyte or Component Names Orotic Acid
Shipping Condition Room Temperature
Molecular Formula C5 H4 N2 O4
Certifications/Compliance ISO 17034
Synonym 4-Pyrimidinecarboxylic acid, 1,2,3,6-tetrahydro-2,6-dioxo-, 2,4-Dioxo-1H-pyrimidine-6-carboxylic acid, Orotic acid (8CI), 1,2,3,6-Tetrahydro-2,6-dioxo-4-pyrimidinecarboxylic acid, 2,4-Dihydroxypyrimidine-6-carboxylic acid, 2,6-Dihydroxy-4-pyrimidinecarboxylic acid, 2,6-Dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid, 6-Carboxyuracil, 6-Uracilcarboxylic acid, Animal galactose factor, NSC 79560, NSC 9791, Orodin, Orotonin, Oroturic, Orotyl, Vitamin B13, Whey factor, Orotic Acid
InChI Formula InChI=1S/C5H4N2O4/c8-3-1-2(4(9)10)6-5(11)7-3/h1H,(H,9,10)(H2,6,7,8,11)
SMILES OC(=O)C1=CC(=O)NC(=O)N1
IUPAC Name 2,4-dioxo-1H-pyrimidine-6-carboxylic acid
Molecular Weight (g/mol) 156.1
Quantity 250 mg
Formula Weight 156.0171 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.