missing translation for 'onlineSavingsMsg'
Learn More

OSMR beta Antibody (469221) [Janelia Fluor™ 669], R&D Systems™

Product Code. 30171122 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30171122 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30171122 Supplier Novus Biologicals Supplier No. FAB4389JF669
  • ONLINE EXCLUSIVE PRICE
    722.31 €
    614.25 € / 0.1mL
    Save 108.06 €
    14% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

OSMR beta Monoclonal specifically detects OSMR beta in Human samples. It is validated for CyTOF, ELISA, Flow Cytometry, Gene Knock-Out, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OSMR beta
Applications CyTOF, ELISA, Flow Cytometry, Gene Knock-Out, Western Blot
Classification Monoclonal
Clone 469221
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias IL-31 receptor subunit beta, IL-31R subunit beta, IL-31RB, IL-31R-beta, Interleukin-31 receptor subunit beta, MGC150627, MGC75127, oncostatin M receptor, oncostatin-M specific receptor beta subunit, oncostatin-M-specific receptor subunit beta, OSMR, OSMRB, OSMRBMGC150626, PLCA1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human OSM Rβ, Glu28-Ser739, Accession # Q99650
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9180
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.