missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OTUD6A Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94731-0.02ml
This item is not returnable.
View return policy
Description
OTUD6A Polyclonal antibody specifically detects OTUD6A in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| OTUD6A | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DUBA2, DUBA-2, OTU domain containing 6A | |
| Recombinant protein of human OTUD6A | |
| 0.02 mL | |
| Cell Biology | |
| 139562 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction