missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OTUD6A Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
232.00 € - 470.00 €
Specifications
| Antigen | OTUD6A |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18642812
|
Novus Biologicals
NBP2-94731-0.02ml |
0.02 mL | |||||||
|
18621512
|
Novus Biologicals
NBP2-94731-0.1ml |
0.1 mL |
470.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OTUD6A Polyclonal antibody specifically detects OTUD6A in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| OTUD6A | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS (pH 7.3), 50% glycerol | |
| 139562 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DUBA2, DUBA-2, OTU domain containing 6A | |
| Recombinant protein of human OTUD6A | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title