missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid Dihydrate, Mikromol™

Product Code. 30051085
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051085 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051085 Supplier Mikromol Supplier No. MM0928.00
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Oxalic Acid Dihydrate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oxalic Acid Dihydrate
CAS 6153-56-6
Analyte or Component Names Oxalic Acid Dihydrate
Shipping Condition Room Temperature
Molecular Formula C22 H24 N2 O9 . 2 H2 O
Certifications/Compliance ISO 9001
Synonym Oxaliplatin Imp. A (EP), Oxaliplatin USP Related Compound A, Oxaliplatin USP RC A, Oxalic Acid Dihydrate, Ethanedioic Acid Dihydrate, Oxaliplatin Imp. A (EP) as Dihydrate, Ethanedioic acid hydrate (1:2), Oxaliplatin Related Compound A as Dihydrate, Ascorbic Acid Impurity E as Dihydrate, Oxaliplatin Impurity A as Dihydrate
SMILES O.O.OC(=O)C(=O)O
Molecular Weight (g/mol) 496.46
Quantity 500 mg
Formula Weight 496.4645 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.