missing translation for 'onlineSavingsMsg'
Learn More

Oxybenzone, TRC

Product Code. 30012019
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012019 10 g 10g
30006856 1 g 1g
30005420 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012019 Supplier Toronto Research Chemicals Supplier No. TRCO86730010G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131-57-7
Molecular Formula C14 H12 O3
Molecular Weight (g/mol) 228.24
Synonym (2-Hydroxy-4-methoxyphenyl)phenylmethanone, 2-Hydroxy-4-methoxybenzophenone, (2-Hydroxy-4-methoxyphenyl)phenylmethanone, 2-Benzoyl-5-methoxyphenol, 4-Methoxy-2-hydroxybenzophenone, ASL 24, Aduvex 24, Advastab 45, Anuvex, Benzophenone 3, Chimassorb 90, Cyasorb UV 9, Cyasorb UV 9 Light Absorber, Escalol 567, Eusolex 4360, Jeescreen Benzophenone 3, Kemisorb 11, MOB, NSC 7778, Neo Heliopan BB, Ongrostab HMB, Onzone, Oxybenzon, Oxybenzone, Seesorb 101, Spectra-Sorb UV 9, Sumisorb 110, Sunscreen UV 15, Syntase 62, Tinosorb B 3, UF 3, UV 9, Uvasorb MET/C, Uvinul 3040, Uvinul 9, Uvinul M 40, Uvistat 24, Viosorb 110
IUPAC Name (2-hydroxy-4-methoxyphenyl)-phenylmethanone
SMILES COc1ccc(C(=O)c2ccccc2)c(O)c1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C14H12O3/c1-17-11-7-8-12(13(15)9-11)14(16)10-5-3-2-4-6-10/h2-9,15H,1H3
Formula Weight 228.0786
Percent Purity >95%
Chemical Name or Material Oxybenzone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.