missing translation for 'onlineSavingsMsg'
Learn More

Oxybutynin Hydrochloride EP Impurity B, Mikromol™

Product Code. 30051054
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051054 25 mg 25mg
17921796 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30051054 Supplier Mikromol Supplier No. MM0047.060025
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Oxybutynin Hydrochloride EP Impurity B
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(Diethylamino)but-2-ynyl 2-Hydroxy-2,2-diphenylacetate (Diphenyl Analogue of Oxybutynin)
CAS 14943-53-4
Analyte or Component Names 4-(Diethylamino)but-2-ynyl 2-Hydroxy-2,2-diphenylacetate (Diphenyl Analogue of Oxybutynin)
Shipping Condition Room Temperature
Molecular Formula C22 H25 N O3
Certifications/Compliance ISO 17025
Synonym Oxybutynin Hydrochloride Imp. B (EP), Oxybutynin Imp. B (EP), 4-(Diethylamino)but-2-ynyl 2-Hydroxy-2,2-diphenylacetate, Diphenyl Analogue of Oxybutynin, Oxybutynin Hydrochloride Impurity B, Oxybutynin Impurity B
InChI Formula InChI=1S/C22H25NO3/c1-3-23(4-2)17-11-12-18-26-21(24)22(25,19-13-7-5-8-14-19)20-15-9-6-10-16-20/h5-10,13-16,25H,3-4,17-18H2,1-2H3
SMILES CCN(CC)CC#CCOC(=O)C(O)(c1ccccc1)c2ccccc2
IUPAC Name 4-(diethylamino)but-2-ynyl 2-hydroxy-2,2-diphenylacetate
Molecular Weight (g/mol) 351.44
Quantity 25 mg
Formula Weight 351.1834 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.