missing translation for 'onlineSavingsMsg'
Learn More

Oxyphenbutazone 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30318730
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30318730 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318730 Supplier Dr. Ehrenstorfer Supplier No. DREA15810000AL1000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oxyphenbutazone
CAS 129-20-4
Analyte or Component Names Oxyphenbutazone
Shipping Condition +20°C
Molecular Formula C19 H20 N2 O3
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym 3,5-Pyrazolidinedione, 4-butyl-1-(p-hydroxyphenyl)-2-phenyl- (6CI,7CI,8CI), 4-Butyl-1-(4-hydroxyphenyl)-2-phenyl-3,5-pyrazolidinedione, 1-(p-Hydroxyphenyl)-2-phenyl-4-butyl-3,5-pyrazolidinedione, 1-Phenyl-2-(p-hydroxyphenyl)-3,5-dioxo-4-butylpyrazolidine, 1-Phenyl-2-(p-hydroxyphenyl)-3,5-dioxo-4-n-butylpyrazolidine, 1-p-Hydroxyphenyl-2-phenyl-3,5-dioxo-4-n-butylpyrazolidine, 4-Butyl-2-(4-hydroxyphenyl)-1-phenyl-3,5-dioxopyrazolidine, 4-Butyl-2-(p-hydroxyphenyl)-1-phenyl-3,4-pyrazolidinedione, 4-Butyl-2-(p-hydroxyphenyl)-1-phenyl-3,5-pyrazolidinedione, Butapirone, Californit, Crovaril, Flamaril, Floghene, Flogitolo, Flogoril, Frabel, G 27202, Metabolite I, NSC 526053, Neo-Farmadol, Offitril, Oxalid, Oxazolidin, Oxyphenbutazone, Oxyphenobutazone, Oxyphenylbutazone, Rapostan, Ro 04-4410, Romaxin, Suganril, Tandacote, Tandearil, Tanderil, Telidal, Tendearil, Visubutina, p-Hydroxyphenylbutazone, p-Oxyphenylbutazone
InChI Formula InChI=1S/C19H20N2O3/c1-2-3-9-17-18(23)20(14-7-5-4-6-8-14)21(19(17)24)15-10-12-16(22)13-11-15/h4-8,10-13,17,22H,2-3,9H2,1H3
SMILES CCCCC1C(=O)N(N(C1=O)c2ccc(O)cc2)c3ccccc3
IUPAC Name 4-butyl-1-(4-hydroxyphenyl)-2-phenylpyrazolidine-3,5-dione
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 324.37
Quantity 1 mL
Formula Weight 324.1474 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.