missing translation for 'onlineSavingsMsg'
Learn More

p-Fluorotoluene, 97%

Product Code. 10409610 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
100 g
250 g
Packaging:
Glass bottle
Unit Size:
100g
10g
250g
This item is not returnable. View return policy

Product Code. 10409610

Brand: Thermo Scientific Acros 119672500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 352-32-9 | C7H7F | 110.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352-32-9
MDL Number MFCD00000358
InChI Key WRWPPGUCZBJXKX-UHFFFAOYSA-N
Synonym 4-fluorotoluene, p-fluorotoluene, benzene, 1-fluoro-4-methyl, p-fluoromethylbenzene, toluene, p-fluoro, 1-methyl-4-fluorobenzene, p-fluoromethyl benzene, unii-nud6wz1b8f, p-tolyl fluoride, para-fluorotoluene
PubChem CID 9603
IUPAC Name 1-fluoro-4-methylbenzene
SMILES CC1=CC=C(C=C1)F

Specifications

Melting Point -56°C
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 116°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C7H7F
Linear Formula CH3C6H4F
Refractive Index 1.4674 to 1.4694
Quantity 250 g
Beilstein 05, 290
Merck Index 15, 4212
Specific Gravity 1
Solubility Information Solubility in water: insoluble.,soluble in alcohols,ether
Molecular Weight (g/mol) 110.13
Formula Weight 110.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material p-Fluorotoluene, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • p-Fluorotoluene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/physician/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.