missing translation for 'onlineSavingsMsg'
Learn More

p21/CIP1/CDKN1A Antibody (CL13081) [CoraFluor™ 1], Novus Biologicals™

Product Code. 30297358 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30297358 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30297358 Supplier Novus Biologicals Supplier No. NBP318554CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

p21/CIP1/CDKN1A Monoclonal antibody specifically detects p21/CIP1/CDKN1A in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen p21/CIP1/CDKN1A
Applications Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone CL13081
Conjugate CoraFluor 1
Formulation PBS
Gene Alias CAP20cyclin-dependent kinase inhibitor 1, CDK-interacting protein 1, CDKN1melanoma differentiation associated protein 6, CIP1WAF1CDK-interaction protein 1, cyclin-dependent kinase inhibitor 1A (p21, Cip1), MDA6, MDA-6, Melanoma differentiation-associated protein 6, p21, p21CIP1, p21Cip1/Waf1, PIC1, SDI1DNA synthesis inhibitor, wild-type p53-activated fragment 1
Host Species Mouse
Immunogen This antibody was developed using a synthetic peptide derived from P38936, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, DNA Repair, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 1026
Target Species Human
Content And Storage Store at 4⪚C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.