missing translation for 'onlineSavingsMsg'
Learn More

P2X2/P2RX2 Antibody [HRP], Novus Biologicals Biologicals™

Product Code. 30511283 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30511283 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30511283 Supplier Novus Biologicals Supplier No. NBP338379H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

P2X2/P2RX2 Polyclonal antibody specifically detects P2X2/P2RX2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen P2X2/P2RX2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias ATP receptor, MGC129601, P2X purinoceptor 2, P2X2P2X Receptor, subunit 2, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 60-320 of human P2X2/P2RX2 (NP_733782.1).,, Sequence:, VFIVQKSYQESETGPESSIITKVKGITTSEHKVWDVEEYVKPPEGGSVFSIITRVEATHSQTQGTCPESIRVHNATCLSDADCVAGELDMLGNGLRTGRCVPYYQGPSKTCEVFGWCPVEDGASVSQFLGTMAPNFTILIKNSIHYPKFHFSKGNIADRTDGYLKRCTFHEASDLYCPIFKLGFIVEKAGESFTELAHKGGVIGVIINWDCDLDLPASECNPKYSFRRLDPKHVPASSGYNFRFAKYYKINGTTTRTLIKA
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 22953
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.