missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y11/P2RY11 Antibody (505214), mFluor Violet 500 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
P2Y11/P2RY11 Monoclonal antibody specifically detects P2Y11/P2RY11 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
Specifications
| Antigen | P2Y11/P2RY11 |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?