missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y12/P2RY12 Antibody (SR1887), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22528-50ul
This item is not returnable.
View return policy
Description
P2Y12/P2RY12 Monoclonal antibody specifically detects P2Y12/P2RY12 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| P2Y12/P2RY12 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunoprecipitation | |
| SR1887 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| ADPG-R, Gi-coupled ADP receptor HORK3, HORK3P2Y12 platelet ADP receptor, P2T(AC), P2Y purinoceptor 12, P2Y(AC), P2Y(ADP), P2Y(cyc), P2Y12ADP-glucose receptor, purinergic receptor P2RY12, purinergic receptor P2Y, G-protein coupled, 12, putative G-protein coupled receptor, SP1999G-protein coupled receptor SP1999 | |
| A synthesized peptide derived from human P2Y12/P2RY12 (Uniprot #: Q9H244) | |
| 50 μg | |
| GPCR | |
| 64805 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction