missing translation for 'onlineSavingsMsg'
Learn More

P2Y12/P2RY12 Antibody (SR1887), Novus Biologicals™

Rabbit Monoclonal Antibody

359.00 € - 449.00 €

Specifications

Antigen P2Y12/P2RY12
Clone SR1887
Dilution Western Blot 1:500-1:2000, Immunoprecipitation 1:50
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18622644
View Documents
Novus Biologicals
NBP3-22528-100ul
100 μg
449.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18646274
View Documents
Novus Biologicals
NBP3-22528-50ul
50 μg
359.00 €
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

P2Y12/P2RY12 Monoclonal antibody specifically detects P2Y12/P2RY12 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications

Specifications

P2Y12/P2RY12
Western Blot 1:500-1:2000, Immunoprecipitation 1:50
Monoclonal
Purified
RUO
Human, Mouse, Rat
ADPG-R, Gi-coupled ADP receptor HORK3, HORK3P2Y12 platelet ADP receptor, P2T(AC), P2Y purinoceptor 12, P2Y(AC), P2Y(ADP), P2Y(cyc), P2Y12ADP-glucose receptor, purinergic receptor P2RY12, purinergic receptor P2Y, G-protein coupled, 12, putative G-protein coupled receptor, SP1999G-protein coupled receptor SP1999
A synthesized peptide derived from human P2Y12/P2RY12 (Uniprot #: Q9H244)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR1887
Western Blot, Immunoprecipitation
Unconjugated
Rabbit
GPCR
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
64805
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.