missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p38 alpha, p Tyr182 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-82097-0.025ML
This item is not returnable.
View return policy
Description
p38 alpha Polyclonal specifically detects p38 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| p38 (p Tyr182) | |
| Polyclonal | |
| Unconjugated | |
| PBS [pH 7.4] containing 150mM NaCl, and 50% glycerol (without Mg2+ and Ca2+) with 0.02% Sodium Azide | |
| CSAID-binding protein, Csaids binding protein, CSBP, CSBP1MAX-interacting protein 2, CSBP2, CSPB1, cytokine suppressive inflammatory drug binding protein, Cytokine suppressive inflammatory drug-binding protein, EC 2.7.11, EC 2.7.11.24, EXIP, MAP kinase 14, MAP kinase MXI2, MAP kinase p38 alpha, MAPK 14, mitogen-activated protein kinase 14, Mitogen-activated protein kinase p38 alpha, Mxi2, p38, p38 MAP kinase, p38 mitogen activated protein kinase, p38ALPHA, p38alpha Exip, PRKM14, PRKM15, RK, SAPK2A, stress-activated protein kinase 2A | |
| Rabbit | |
| 43 kDa | |
| 0.025 mL | |
| Cancer, Cell Cycle and Replication, Immunology, Innate Immunity, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Signal Transduction | |
| 1432 | |
| Rat, Human, Mouse | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
| Q16539 | |
| MAPK14 | |
| The antiserum was produced against synthesized phosphopeptide derived from human p38 MAPK around the phosphorylation site of tyrosine 182 (T-G-YP-V-A). | |
| Affinity Purified | |
| RUO | |
| Primary | |
| p38 MAPK (specific to Phospho-Tyr182) detects endogenous levels of P38MAPK only when phosphorylated at tyrosine 182. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction