missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p38 alpha, p Tyr182 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
255.00 € - 397.00 €
Specifications
| Antigen | p38 (p Tyr182) |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18243673
|
Novus Biologicals
NB100-82097-0.025ML |
0.025 mL |
255.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18746203
|
Novus Biologicals
NB100-82097 |
0.05 mL |
397.00 €
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p38 alpha Polyclonal specifically detects p38 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| p38 (p Tyr182) | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Immunology, Innate Immunity, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Signal Transduction | |
| Q16539 | |
| 1432 | |
| IgG | |
| Affinity Purified | |
| 43 kDa |
| 1.0 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Rat, Human, Mouse | |
| CSAID-binding protein, Csaids binding protein, CSBP, CSBP1MAX-interacting protein 2, CSBP2, CSPB1, cytokine suppressive inflammatory drug binding protein, Cytokine suppressive inflammatory drug-binding protein, EC 2.7.11, EC 2.7.11.24, EXIP, MAP kinase 14, MAP kinase MXI2, MAP kinase p38 alpha, MAPK 14, mitogen-activated protein kinase 14, Mitogen-activated protein kinase p38 alpha, Mxi2, p38, p38 MAP kinase, p38 mitogen activated protein kinase, p38ALPHA, p38alpha Exip, PRKM14, PRKM15, RK, SAPK2A, stress-activated protein kinase 2A | |
| MAPK14 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title