missing translation for 'onlineSavingsMsg'
Learn More

p53 Ab-4, Mouse Monoclonal Antibody,Lab Vision™ Epredia™

Achieve accurate, reproducible results with the Epredia™ p53 Ab-4, Mouse Monoclonal Antibody.

Brand:  Epredia MS-182-P1

 View more versions of this product

Product Code. 12660227

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Beskrivelse

Beskrivelse

p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein p53 senses DNA damage and possibly facilitating repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.

Host Species: Mouse

Clone: PAb 122

Isotype: IgG2b/κ

Species Reactivity: Human, Monkey, Mouse, Rat, and Hamster. Others not known.

Epitope: aa 370-378

Immunogen: SV40-transformed mouse B4 cells

Molecular Weight: 53kDa

Positive Control: MDA-231 cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured verified)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Tekniske data

Tekniske data

p53 Ab-4
Monoclonal
200μg/mL
Mouse
500 μL
Cancer and Tumor Biology
Hamster, Human, Mouse, Non-human Primate, Rat
IgG2b κ
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot
PAb 122
Unconjugated
SV40-transformed mouse B4 cells
RUO
Primary
Concentrated
Produktforslag

Produktforslag

Videoer
SDS
Dokumenter

Dokumenter

Certifikater
Kampagner

Kampagner

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.