Learn More
p53 Ab-4, Mouse Monoclonal Antibody,Lab Vision™ Epredia™
Achieve accurate, reproducible results with the Epredia™ p53 Ab-4, Mouse Monoclonal Antibody.
129.00 € - 560.00 €
Specifications
| Antigen | p53 Ab-4 |
|---|---|
| Clone | PAb 122 |
| Applications | Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Concentration | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Concentration | Price | Quantity & Availability | ||||
|
12650227
|
Epredia
MS-182-P0 |
100 μL | 200μg/mL | ||||||
|
12660227
|
Epredia
MS-182-P1 |
500 μL | 200μg/mL | ||||||
|
12670227
|
Epredia
MS-182-P1ABX |
100 μL | 1mg/mL | ||||||
|
12680227
|
Epredia
MS-182-PABX |
200 μL | 1mg/mL | ||||||
Description
p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein p53 senses DNA damage and possibly facilitating repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.Host Species: Mouse
Clone: PAb 122
Isotype: IgG2b/κ
Species Reactivity: Human, Monkey, Mouse, Rat, and Hamster. Others not known.
Epitope: aa 370-378
Immunogen: SV40-transformed mouse B4 cells
Molecular Weight: 53kDa
Positive Control: MDA-231 cells
Cellular Localization: Nuclear
Recommended for:
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Denatured verified)
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
| p53 Ab-4 | |
| Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Cancer and Tumor Biology | |
| SV40-transformed mouse B4 cells | |
| Primary |
| PAb 122 | |
| Monoclonal | |
| Concentrated | |
| RUO | |
| Hamster, Human, Mouse, Non-human Primate, Rat | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.