missing translation for 'onlineSavingsMsg'
Learn More

p53 Ab-4, Mouse Monoclonal Antibody,Lab Vision™ Epredia™

Achieve accurate, reproducible results with the Epredia™ p53 Ab-4, Mouse Monoclonal Antibody.

Brand:  Epredia MS-182-PABX

 View more versions of this product

Product Code. 12680227

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein p53 senses DNA damage and possibly facilitating repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.

Host Species: Mouse

Clone: PAb 122

Isotype: IgG2b/κ

Species Reactivity: Human, Monkey, Mouse, Rat, and Hamster. Others not known.

Epitope: aa 370-378

Immunogen: SV40-transformed mouse B4 cells

Molecular Weight: 53kDa

Positive Control: MDA-231 cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured verified)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

p53 Ab-4
Monoclonal
1mg/mL
Mouse
200 μL
Cancer and Tumor Biology
Hamster, Human, Mouse, Non-human Primate, Rat
IgG2b κ
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot
PAb 122
Unconjugated
SV40-transformed mouse B4 cells
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.