Learn More
p73 Antibody (5B1288) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 28 publications
Brand:  Novus Biologicals NB100-56674SS
 View more versions of this product
Product Code. 18070074
  • 235.00 € / 25µg
Description
p73 Monoclonal specifically detects p73 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
p73
Monoclonal
1.0 mg/mL
Western Blot 1-3 ug/ml, Chromatin Immunoprecipitation 1:20 - 1:1000. Use reported in scientific literature (Accardi et al), Immunohistochemistry 1:100 - 1:200. Use reported in scientific literature (PMID 19816568), Immunocytochemistry/Immunofluorescence 1:10-1:2000. Use reported in scientific literature (Sayan et al), Immunoprecipitation 1:20 - 1:1000. Use reported in scientific literature (Sayan et al), Immunohistochemistry-Paraffin 1:100 - 1:200, Chromatin Immunoprecipitation (ChIP) 1:20-1:1000
p53-related protein, P73p53-like transcription factor, TA p73, TAp73, tumor protein p73
Mouse
73 kDa
0.025 mg
Apoptosis, Cancer, Tumor Suppressors
7161
Human, Mouse
Purified
Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
5B1288
Unconjugated
O15350
TP73
This antibody was raised against full-length human p73. The epitope is thought to lie around the center of the molecule (NP_005418).
Protein G purified
RUO
Primary
It reacts with alpha, beta, gamma and delta isoforms of mouse and human p73 as well as the dominant negative p73 (Costanzo, et al, 2002). Because of its reactivity pattern it may be regarded as anti-pan p73. NB100-56674 has been shown to recognize all the known alternative splicing variants of human and mouse p73 (Costanzo et al, 2002). The antibody does not cross react with p53.
Store at -20C. Avoid freeze-thaw cycles.
IgG1 κ