missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p73 Antibody (5B1288) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 28 publications
235.00 € - 550.00 €
Specifications
| Antigen | p73 |
|---|---|
| Clone | 5B1288 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1-3 ug/ml, Chromatin Immunoprecipitation 1:20 - 1:1000. Use reported in scientific literature (Accardi et al), Immunohistochemistry 1:100 - 1:200. Use reported in scientific literature (PMID 19816568), Immunocytochemistry/Immunofluorescence 1:10-1:2000. Use reported in scientific literature (Sayan et al), Immunoprecipitation 1:20 - 1:1000. Use reported in scientific literature (Sayan et al), Immunohistochemistry-Paraffin 1:100 - 1:200, Chromatin Immunoprecipitation (ChIP) 1:20-1:1000 |
| Applications | Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18070074
|
Novus Biologicals
NB100-56674SS |
0.025 mg |
235.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18710424
|
Novus Biologicals
NB100-56674 |
0.1 mg |
550.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p73 Monoclonal specifically detects p73 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| p73 | |
| 1.0 mg/mL | |
| Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Tumor Suppressors | |
| O15350 | |
| 7161 | |
| This antibody was raised against full-length human p73. The epitope is thought to lie around the center of the molecule (NP_005418). | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| 73 kDa |
| 5B1288 | |
| Western Blot 1-3 ug/ml, Chromatin Immunoprecipitation 1:20 - 1:1000. Use reported in scientific literature (Accardi et al), Immunohistochemistry 1:100 - 1:200. Use reported in scientific literature (PMID 19816568), Immunocytochemistry/Immunofluorescence 1:10-1:2000. Use reported in scientific literature (Sayan et al), Immunoprecipitation 1:20 - 1:1000. Use reported in scientific literature (Sayan et al), Immunohistochemistry-Paraffin 1:100 - 1:200, Chromatin Immunoprecipitation (ChIP) 1:20-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| p53-related protein, P73p53-like transcription factor, TA p73, TAp73, tumor protein p73 | |
| TP73 | |
| IgG1 κ | |
| Protein G purified | |
| It reacts with alpha, beta, gamma and delta isoforms of mouse and human p73 as well as the dominant negative p73 (Costanzo, et al, 2002). Because of its reactivity pattern it may be regarded as anti-pan p73. NB100-56674 has been shown to recognize all the known alternative splicing variants of human and mouse p73 (Costanzo et al, 2002). The antibody does not cross react with p53. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title