missing translation for 'onlineSavingsMsg'
Learn More

PA28 Activator beta Subunit/PSME2 Antibody [mFluor Violet 610 SE], Novus Biologicals Biologicals™

Product Code. 30490424 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30490424 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30490424 Supplier Novus Biologicals Supplier No. NBP335372MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA28 Activator beta Subunit/PSME2 Polyclonal antibody specifically detects PA28 Activator beta Subunit/PSME2 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PA28 Activator beta Subunit/PSME2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias 11S regulator complex beta subunit, 11S regulator complex subunit beta, Activator of multicatalytic protease subunit 2, MCP activator, 31-kD subunit, PA28b, PA28betacell migration-inducing protein 22, proteasome (prosome, macropain) activator subunit 2 (PA28 beta), Proteasome activator 28 subunit beta, proteasome activator 28-beta, proteasome activator complex subunit 2, proteasome activator hPA28 subunit beta, REGbeta, REG-beta
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-239 of human PA28 Activator beta Subunit/PSME2 (NP_002809.2).,, Sequence:, MAKPCGVRLSGEARKQVEVFRQNLFQEAEEFLYRFLPQKIIYLNQLLQEDSLNVADLTSLRAPLDIPIPDPPPKDDEMETDKQEKKEVHKCGFLPGNEKVLSLLALVKPEVWTLKEKCILVITWIQHLIPKIEDGNDFGVAIQEKVLERVNAVKTKVEAFQTTISKYFSERGDAVAKASKETHVMDYRALVHERDEAAYGELRAMVLDLRAFYAELYHIISSNLEKIVNPKGEEKPSMY
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5721
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.