missing translation for 'onlineSavingsMsg'
Learn More

PACSIN3 Antibody [Alexa Fluor« 488], Novus Biologicals Biologicals™

Product Code. 30516789 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30516789 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30516789 Supplier Novus Biologicals Supplier No. NBP338573AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PACSIN3 Polyclonal antibody specifically detects PACSIN3 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PACSIN3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias Endophilin I, FLJ17861, protein kinase C and casein kinase substrate in neurons 3, protein kinase C and casein kinase substrate in neurons protein 3, SDPIII, SH3 domain-containing protein 6511, syndapin III
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 175-424 of human PACSIN3 (NP_057307.2).,, Sequence:, KADSAVSQEQLRKLQERVERCAKEAEKTKAQYEQTLAELHRYTPRYMEDMEQAFETCQAAERQRLLFFKDMLLTLHQHLDLSSSEKFHELHRDLHQGIEAASDEEDLRWWRSTHGPGMAMNWPQFEEWSLDTQRTISRKEKGGRSPDEVTLTSIVPTRDGTAPPPQSPGSPGTGQDEEWSDEESPRKAATGVRVRALYDYAGQEADELSFRAGEELLKMSEEDEQGWCQGQLQSGRIGLYPANYVECVGA
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Diabetes Research, Neuroscience, Protein Kinase, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 29763
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.