missing translation for 'onlineSavingsMsg'
Learn More

Pamaquine Naphthoate, LoGiCal

Product Code. 30458960
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30458960 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458960 Supplier LoGiCal Supplier No. LGCFOR1437.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pamaquine Naphthoate
CAS 635-05-2
Analyte or Component Names Pamaquine Naphthoate
Shipping Condition Room Temperature
Molecular Formula C23 H16 O6 . C19 H29 N3 O
Certifications/Compliance ISO 9001
Synonym 8-[[4-(diethylamino)-1-methylbutyl]amino]-6-methoxyquinoline, 4,4'-methylenebis[3-hydroxy-2-naphthoic acid], N1,N1-diethyl-N4-(6-methoxy-8-quinolinyl)-1,4-pentanediamine 4,4'-methylenebis[3-hydroxy-2-naphthalenecarboxylate], 8-[[4-(diethylamino)-1-methylbutyl]amino]-6-methoxy-quinoline, 4,4'-methylenebis[3-hydroxy-2-naphthoate], 2,2'-dihydroxy-1,1'-dinaphthylmethane-3,3'-dicarboxylic acid 6-methoxy-8-(4-diethylamino-1-methylbutyl)-aminoquinoline salt, 4,4'-8-[4-diethylamino-1-methylbutyl)amino]-6-methoxyquinoline methylenebis(3-hydroxy-2-naphthoic acid) salt, Aminoquin naphthoate, Beprochine naphthoate, Gamefar naphthoate, NSC 14220, Pamaquine embonate, Pamaquine naphthoate, Pamaquine pamoate, Pamoquine ebonate, Plasmochin naphthoate, Plasmoquine naphthoate, Praequine naphthoate, Quipenyl naphthoate
InChI Formula InChI=1S/C23H16O6.C19H29N3O/c24-20-16(14-7-3-1-5-12(14)9-18(20)22(26)27)11-17-15-8-4-2-6-13(15)10-19(21(17)25)23(28)29;1-5-22(6-2)12-8-9-15(3)21-18-14-17(23-4)13-16-10-7-11-20-19(16)18/h1-10,24-25H,11H2,(H,26,27)(H,28,29);7,10-11,13-15,21H,5-6,8-9,12H2,1-4H3
Solution Type Neat
SMILES CCN(CC)CCCC(C)Nc1cc(OC)cc2cccnc12.OC(=O)c3cc4ccccc4c(Cc5c(O)c(cc6ccccc56)C(=O)O)c3O
IUPAC Name 4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid;1-N,1-N-diethyl-4-N-(6-methoxyquinolin-8-yl)pentane-1,4-diamine
Industry Type Clinical Testing
Molecular Weight (g/mol) 703.82
Quantity 5 mg
Formula Weight 703.3258 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.