missing translation for 'onlineSavingsMsg'
Learn More

Pancreatic Amylase Alpha Antibody (1C7) [DyLight 488], Novus Biologicals Biologicals™

Product Code. 30518302 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30518302 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30518302 Supplier Novus Biologicals Supplier No. NBP342927G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Pancreatic Amylase Alpha Monoclonal antibody specifically detects Pancreatic Amylase Alpha in Human,Porcine,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pancreatic Amylase Alpha
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 1C7
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias AMY2, AMY2A amylase, alpha 2A (pancreatic), AMY2B, PA, pancreatic alpha-amylase
Host Species Mouse
Immunogen amylase, alpha 2A(pancreatic) - (Uniprot# P04746)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 279
Target Species Human, Pig, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.