Learn More
Panendothelial Cell Antigen Antibody (MECA-32), Alexa Fluor™ 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals NB100-77668AF488SS
306.00 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
 View more versions of this product
Product Code. 18711510
  • 382.00 € / 0.10mL
Description
Panendothelial Cell Antigen Monoclonal antibody specifically detects Panendothelial Cell Antigen in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Panendothelial Cell Antigen
Monoclonal
Alexa Fluor 488
FELS, Fenestrated endothelial-linked structure protein, gp68, MECA32, MECA-32, Panendothelial Cell Antigen, plasmalemma vesicle associated protein, Plasmalemma Vesicle Protein 1, PV-1 protein
Mouse lymph node stromal cells.
0.1 mL
Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers
84094
Store at 4°C in the dark.
IgG2a κ
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
MECA-32
50mM Sodium Borate
Rat
Protein G purified
RUO
Primary
Human, Mouse
Purified