missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Panendothelial Cell Antigen Antibody (MECA-32), Alexa Fluor™ 488, Novus Biologicals™
Rat Monoclonal Antibody
382.00 € - 581.00 €
Specifications
| Antigen | Panendothelial Cell Antigen |
|---|---|
| Clone | MECA-32 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Conjugate | Alexa Fluor 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18722853
|
Novus Biologicals
NB100-77668AF488 |
0.5 mL |
581.00 €
0.50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18711510
|
Novus Biologicals
NB100-77668AF488SS |
0.1 mL |
382.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Panendothelial Cell Antigen Monoclonal antibody specifically detects Panendothelial Cell Antigen in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOFSpecifications
| Panendothelial Cell Antigen | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| Alexa Fluor 488 | |
| Rat | |
| Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 50mM Sodium Borate | |
| 84094 | |
| IgG2a κ | |
| Protein G purified |
| MECA-32 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| FELS, Fenestrated endothelial-linked structure protein, gp68, MECA32, MECA-32, Panendothelial Cell Antigen, plasmalemma vesicle associated protein, Plasmalemma Vesicle Protein 1, PV-1 protein | |
| Mouse lymph node stromal cells. | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title